Skip to Content
Merck
All Photos(1)

Key Documents

ABT165

Sigma-Aldrich

Anti-RAB8A Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Ras-related protein Rab-8A, Oncogene c-mel

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

species reactivity (predicted by homology)

mouse (based on 100% sequence homology), bovine (based on 100% sequence homology)

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... RAB8A(4218)
mouse ... Rab8A(17274)

General description

Ras-related protein Rab-8A (RAB8A) is one of two Rab8 isoforms, which belong to the superfamily of GTPases. They cycle between an active GTP-bound state, formed from interaction with guanine nucleotide exchange factors (GEFs) such as Rabin8 and GRAB; and an inactive GDP-bound state via interaction with GTPase activating proteins (GAPs) such as TBC1D1. Rab8A activates various effector molecules, which are coupled to intracellular signaling networks that mediate membrane and vesicular trafficking, and actin remodeling. Rab8A is found in the golgi network, perinuclear region, plasma membrane, lamellipodia, and cilia, among other regions.

Specificity

This antibody recognizes the C-terminus of RAB8A.

Immunogen

Epitope: C-terminus
KLH-conjugated linear peptide corresponding to the C-terminus of human RAB8A.

Application

Research Category
Cell Structure
Research Sub Category
Vesicular Trafficking
This Anti-RAB8A Antibody is validated for use in Western Blotting for the detection of RAB8A.
Western Blot Analysis: A representative lot from an independent laboratory detected RAB8A in human cerebellum and brain.

Quality

Evaluated by Western Blot in serum starved HeLa cell lysate.

Western Blot Analysis: 1 µg/mL of this antibody detected RAB8A in serum starved HeLa cell lysate.

Target description

~24 kDa observed. Uncharacterized bands may be observed at ~110 kDa and ~38 kDa

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
Serum starved HeLa cell lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service