Skip to Content
Merck
All Photos(4)

Documents

ABE1062

Sigma-Aldrich

Anti-monomethyl Histone H2A.Z Antibody (Lys 7)

from rabbit, purified by affinity chromatography

Synonym(s):

Histone H2A.Z, H2A/z

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

technique(s)

dot blot: suitable
immunocytochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

monomethylation (Lys 7)

Gene Information

human ... H2AFZ(3015)

General description

H2A.Z, also known as H2AFZ, is a variant of histone H2A that is highly conserved from yeast to humans. Like other histone variants, H2A.Z is encoded by a unique gene, resulting in a slightly divergent type of H2A with specialized functions. H2A.Z is involved in chromatin compaction by stabilizing the histone octamer within the nucleosome. It promotes gene expression through the creation of an open domain of chromatin that is more easily transcribed. H2A.Z is nonrandomly distributed throughout the genome, targeted primarily to pericentric heterochromatin, but excluded from the inactive X chromosome and the nucleolus. H2A.Z is associated with both euchromatin and facultative heterochromatin. DNA methylation can influence chromatin structure and effect gene silencing by excluding H2A.Z, and likewise, H2A.Z protects genes from DNA methylation. Thus, H2A.Z plays a role in regulating heterochromatin silencing and is involved in transcriptional control.

Immunogen

Epitope: Near N-terminal
KLH-conjugated linear peptide corresponding to human monomethyl Histone H2A.Z (Lys 7) near the N-terminal.

Application

Anti-monomethyl Histone H2A.Z (Lys 7) is an antibody against monomethyl Histone H2A.Z (Lys 7) for use in WB, ICC, DB, IP.
Immunocytochemistry Analysis: A 1:2,000 dilution from a representative lot detected monomethyl Histone H2A.Z (Lys 7) in HeLa and NIH/3T3 cells.
Dot Blot Analysis: A 1:1,000 dilution from a representative lot detected monomethyl Histone H2A.Z (Lys 7) in Histone peptides with various modifications.
Immunoprecipitation Analysis: 10 µg from a representative lot immunoprecipitated monomethyl Histone H2A.Z (Lys 7) in 500 µg of HeLa acid extract.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Nuclear Receptors

Quality

Evaluated by Western Blotting in HeLa acid extract.

Western Blotting Analysis: 1.0 µg/mL of this antibody detected monomethyl Histone H2A.Z (Lys 7) in 10 µg of HeLa acid extract.

Target description

~16 kDa observed

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service