Skip to Content
Merck
All Photos(1)

Documents

AB5436P

Sigma-Aldrich

Anti-Brain and Muscle ARNT-Like 1 Antibody

Chemicon®, from rabbit

Synonym(s):

BMAL1, MOP3/CYCLE

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

Drosophila

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
western blot: suitable

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

fruit fly ... Cyc(40162)

Specificity

Recognizes drosophila BMAL1. The immunogen shows no significant sequence homology with mammalian Clock or Per proteins. Significant homology is seen with mammalian USF′s (Upstream Stimulatory Factors 1 and 2) and human and mouse BMAL1 and ARNTs.

SPECIES REACTIVITIES: The immunogen sequence has no homology with mammalian Clock or Per proteins. Reactivity with other species has not been confirmed.

Immunogen

A 22 amino acid peptide of drosophila BMAL1 (Darlington et al. 1998; Rutila et al. 1998; Bae et al. 1998). Gene accession # 3219728.

Application

Anti-Brain & Muscle ARNT-Like 1 Antibody is a Rabbit Polyclonal Antibody for detection of Brain & Muscle ARNT-Like 1 also known as BMAL1, MOP3/CYCLE & has been validated in ELISA & WB.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Transcription Factors
Western blot: 1-10 μg/mL using ECL.

ELISA: 0.5-1 μg/mL using 1 μg/mL control peptide per well.

Optimal working dilutions must be determined by the end user.

Physical form

Affinity purified immunoglobulin. Liquid in phosphate buffered saline, pH 7.4, containing 0.1% BSA and 0.1% sodium azide.

Storage and Stability

Maintain at -20°C in undiluted aliquots for up to 6 months. Avoid repeated freeze/thaw cycles.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service