8.53042
Boc-Tyr(2,6-di-Cl-Bzl)-OH
Novabiochem®
Synonym(s):
Boc-Tyr(2,6-di-Cl-Bzl)-OH, N-α-t.-Boc-O-2,6-dichlorobenzyl-L-tyrosine
About This Item
Recommended Products
Quality Level
product line
Novabiochem®
Assay
≥98% (TLC)
form
powder
reaction suitability
reaction type: Boc solid-phase peptide synthesis
manufacturer/tradename
Novabiochem®
application(s)
peptide synthesis
functional group
hydroxyl
storage temp.
2-30°C
InChI
1S/C21H23Cl2NO5/c1-21(2,3)29-20(27)24-18(19(25)26)11-13-7-9-14(10-8-13)28-12-15-16(22)5-4-6-17(15)23/h4-10,18H,11-12H2,1-3H3,(H,24,27)(H,25,26)
InChI key
DODHIGHXRDNRPP-UHFFFAOYSA-N
Related Categories
General description
Associated Protocols and Technical Articles
Cleavage and Deprotection Protocols for Boc SPPS
Linkage
Analysis Note
Appearance of substance (visual): powder
Identity (IR): conforms
Optical rotation α 25/D (c=2 in ethanol): +17.5 - +22.5 °
Purity (TLC(011A)): ≥ 98 %
Purity (TLC(0811)): ≥ 98 %
Solubility (1 mmole in 2 ml DMF): clearly soluble
To see the solvent systems used for TLC of Novabiochem® products please click here.
Legal Information
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Protocols
Anhydrous HF is the preferred reagent for peptide cleavage from Boc-based resins, versatile and effective for various peptide synthesis.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service