Skip to Content
Merck
All Photos(1)

Key Documents

840086

Sigma-Aldrich

(2-Methoxyethyl) acrylate

(stabilised with hydroquinone monomethyl ether) for synthesis

Synonym(s):

(2-Methoxyethyl) acrylate, Acrylic acid 2-methoxyethyl ester

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
130.14
MDL number:
UNSPSC Code:
12352108
EC Index Number:
221-499-3

vapor pressure

3 hPa ( 20 °C)

Quality Level

form

liquid

autoignition temp.

402 °C

potency

397 mg/kg LD50, oral (Rat)
248 mg/kg LD50, skin (Rabbit)

expl. lim.

1.6 % (v/v)

bp

164 °C/1013 hPa

mp

<-30 °C

transition temp

flash point 56 °C

solubility

116 g/L

density

1.008 g/cm3 at 20 °C

shipped in

ambient

storage temp.

2-8°C

InChI

1S/C6H10O3/c1-3-6(7)9-5-4-8-2/h3H,1,4-5H2,2H3

InChI key

HFCUBKYHMMPGBY-UHFFFAOYSA-N

General description

(stabilised with hydroquinone monomethyl ether) for synthesis

Application

(2-Methoxyethyl) acrylate (stabilised with hydroquinone monomethyl ether) for synthesis. CAS 3121-61-7.

Analysis Note

Assay (GC, area%)≥ 99.0 % (a/a)Density (d 20 °C/ 4 °C)1.009 - 1.013Identity (IR)passes test

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Flam. Liq. 3 - Repr. 1B - Skin Corr. 1C - Skin Sens. 1 - STOT RE 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(C)

60 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service