Skip to Content
Merck
All Photos(1)

Key Documents

8.18395

Sigma-Aldrich

2-Acetylcyclopentanone

for synthesis

Synonym(s):

2-Acetylcyclopentanone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H10O2
CAS Number:
Molecular Weight:
126.15
MDL number:
UNSPSC Code:
12352115
EC Index Number:
216-797-5

Quality Level

Assay

≥98.0% (GC)

form

liquid

bp

72-75 °C/10 hPa

transition temp

flash point 72 °C

density

1.06 g/cm3 at 20 °C

storage temp.

2-8°C

InChI

1S/C7H10O2/c1-5(8)6-3-2-4-7(6)9/h6H,2-4H2,1H3

InChI key

OSWDNIFICGLKEE-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Density (d 20 °C/ 4 °C): 1.061 - 1.062
Identity (IR): passes test

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(C)

72 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service