Skip to Content
Merck
All Photos(1)

Key Documents

8.10560

Sigma-Aldrich

Ruthenium(IV) oxide hydrate (60% Ru)

Synonym(s):

Ruthenium(IV) oxide hydrate (60% Ru)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
O2Ru · xH2O
CAS Number:
Molecular Weight:
133.07 (anhydrous basis)
MDL number:
UNSPSC Code:
12352303
Pricing and availability is not currently available.

form

powder

Quality Level

potency

4580 mg/kg LD50, oral (Rat)

reaction suitability

reagent type: oxidant

storage temp.

2-30°C

Analysis Note

Assay (Ru): ≥ 60.0 %
Identity (IR): passes test

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service