Skip to Content
Merck
All Photos(1)

Key Documents

8.00252

Sigma-Aldrich

Cyanoacetic acid

for synthesis

Synonym(s):

Cyanoacetic acid, Malonic acid mononitrile

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H3NO2
CAS Number:
Molecular Weight:
85.06
MDL number:
UNSPSC Code:
12352106
EC Index Number:
206-743-9
Pricing and availability is not currently available.

vapor pressure

0.1 hPa ( 100 °C)

Assay

≥98.0% (acidimetric)

form

crystals

potency

1500 mg/kg LD50, oral (Rat)
>2000 mg/kg LD50, skin (Rabbit)

pH

1.6 (20 °C, 50 g/L in H2O)

bp

108 °C/20 hPa

mp

65-68 °C

transition temp

flash point 107 °C

solubility

1000 g/L

bulk density

700 kg/m3

storage temp.

2-30°C

InChI

1S/C3H3NO2/c4-2-1-3(5)6/h1H2,(H,5,6)

InChI key

MLIREBYILWEBDM-UHFFFAOYSA-N

Analysis Note

Assay (acidimetric): ≥ 98.0 %
Melting range (lower value): ≥ 66 °C
Melting range (upper value): ≤ 70 °C
Water (K. F.): ≤ 0.60 %
Identity (IR): passes test

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1B

Supplementary Hazards

Storage Class Code

8A - Combustible, corrosive hazardous materials

WGK

WGK 1

Flash Point(C)

107 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service