Skip to Content
Merck
All Photos(1)

Key Documents

681677

Sigma-Aldrich

Wnt Pathway Inhibitor XII

The Wnt Pathway Inhibitor XII controls the biological activity of Wnt Pathway. This small molecule/inhibitor is primarily used for Cancer applications.

Synonym(s):

Wnt Pathway Inhibitor XII, (E)-4-(2,6-Difluorostyryl)-N,N-dimethylaniline, FIDAS3, MAT2A Inhibitor I, Methionine S-Adenosyltransferase 2A Inhibitor I

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H15F2N
CAS Number:
Molecular Weight:
259.29
UNSPSC Code:
12352200

Quality Level

Assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

off-white to pale yellow

solubility

DMSO: 50 mg/mL

shipped in

ambient

storage temp.

2-8°C

General description

A cell-permeable fluorinated N,N-dialkylaminostilbene (FIDAS) compound that inhibits MAT2A-catalyzed SAM (S-adenosylmethionine) synthesis (IC50 =4.9 µM; [L-Met] = [ATP] = 1 mM, [MAT2A] = 10 µg/mL; 20 min preincubation prior to 30 min reaction) by targeting SAM-binding pocket at the MAT2A dimer interface. Shown to downregulate intracellular SAM level (by 64%; 36 h;10 µM) and Wnt signaling in LS174 CRC (colorectal cancer) cultures and is at least 10-fold more potent than its analogs Pterostilbene (Cat. No. 523310) and Resveratrol (Cat. No. 554325) in inhibiting LS174 proliferation both in vitro (by 70%, 85%, and 90% in 4 d, respectively, with 0.1, 0.3, and 1 µM inhibitor) and in mice in vivo (by 75% on d26; 20 mg/kg/day; i.p.).
A cell-permeable fluorinated N,N-dialkylaminostilbene (FIDAS) compound that inhibits MAT2A-catalyzed SAM (S-adenosylmethionine) synthesis by targeting SAM-binding pocket at the MAT2A dimer interface. Shown to downregulate intracellular SAM level (by 64%; 36 h;10 µM) and Wnt signaling in LS174 colorectal cancer cultures and is at least 10-fold more potent than Pterostilbene (Cat. No. 523310) and Resveratrol (Cat. No. 554325) in inhibiting LS174 proliferation both in vitro (by 90% in 4 d; 1 µM) and in mice in vivo (by 75% on d26; 20 mg/kg/day; i.p.).

Packaging

Packaged under inert gas

Warning

Toxicity: Regulatory Review (Z)

Other Notes

Zhang, W., et al. 2013. ACS Chem.Biol.18, In press.
Zhang, W., et al. 2011. J. Med. Chem.54, 7579.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service