65042-M
Wright Giemsa Stain Pack
HARLECO® Designed for use with the Ames Hema-Tek Stainer
Synonym(s):
Hematology Stain Hematrek, Hematology Solutions
About This Item
Recommended Products
Quality Level
transition temp
flash point 11.1 °C
λmax
520-530 nm (Eosin)
645-655 nm (Azure)
application(s)
diagnostic assay manufacturing
hematology
histology
storage temp.
15-25°C
Application
For In-Vitro Diagnostic Use.
For Professional Use.
Designed for use with the Ames Hema-Tek Stainer.
Legal Information
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1
Target Organs
Eyes,Central nervous system
Storage Class Code
3 - Flammable liquids
Flash Point(F)
53.6 °F
Flash Point(C)
12 °C
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service