Skip to Content
Merck
All Photos(1)

Key Documents

6.18507

Supelco

Octane-1-sulfonic acid sodium salt

for ion pair chromatography, LiChropur

Synonym(s):

Octane-1-sulfonic acid sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H17NaO3S
CAS Number:
Molecular Weight:
216.27
MDL number:
UNSPSC Code:
12161902
Pricing and availability is not currently available.

product line

LiChropur

SMILES string

[Na+].[S](=O)(=O)([O-])CCCCCCCC

InChI

1S/C8H18O3S.Na/c1-2-3-4-5-6-7-8-12(9,10)11;/h2-8H2,1H3,(H,9,10,11);/q;+1/p-1

InChI key

HRQDCDQDOPSGBR-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application


  • Determination of meglumine in pharmaceutical formulations using high performance liquid chromatography.: This research presents a method for determining meglumine in pharmaceutical formulations utilizing high performance liquid chromatography (HPLC). Octane-1-sulfonic acid sodium salt is employed as an ion-pairing agent to improve the separation and detection of meglumine. The study demonstrates the effectiveness and reliability of this approach in pharmaceutical analysis, providing valuable insights into its application in quality control and formulation analysis (Jasprica et al., 2011).


Legal Information

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service