Skip to Content
Merck
All Photos(1)

Key Documents

565777

Sigma-Aldrich

γ-Secretase Inhibitor XVI

The γ-Secretase Inhibitor XVI, also referenced under CAS 208255-51-0, controls the biological activity of γ-Secretase. This small molecule/inhibitor is primarily used for Neuroscience applications.

Synonym(s):

γ-Secretase Inhibitor XVI, DAPM, N-[N-3,5-Difluorophenacetyl]-L-alanyl-S-phenylglycine Methyl Ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H20F2N2O4
CAS Number:
Molecular Weight:
390.38
UNSPSC Code:
12352200

Quality Level

Assay

≥97% (HPLC)

form

solid

potency

10 nM IC50

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

solubility

DMSO: 2 mg/mL

shipped in

ambient

storage temp.

2-8°C

InChI

1S/C20H20F2N2O4/c1-12(23-17(25)10-13-8-15(21)11-16(22)9-13)19(26)24-18(20(27)28-2)14-6-4-3-5-7-14/h3-9,11-12,18H,10H2,1-2H3,(H,23,25)(H,24,26)/t12-,18-/m0/s1

InChI key

XAPDRNSTUYCSQC-SGTLLEGYSA-N

General description

A cell-permeable γ-secretase inhibitor that inhibits Aβ secretion (IC50 ~10 nM) by 7PA2 cells (CHO cell line stably transfected with APP751 containing V717F familial Alzheimer′s disease mutation) in a dose-dependent manner. Also exhibits anti-aggregation properties by selectively blocking Aβ dimer and trimer formation and subsequently preventing early Aβ oligomerization.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
γ-secretase Aβ₄₀ in 7PA2 cells
Product does not compete with ATP.
Reversible: no

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 1 month at -20°C.

Other Notes

Walsh, D.M., et al. 2002. Nature 416, 535.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service