552095
R-Spondin 4, Mouse, Recombinant
Synonym(s):
Cristin 4
About This Item
Recommended Products
Assay
≥95% (SDS-PAGE)
Quality Level
form
solid
manufacturer/tradename
Calbiochem®
storage condition
OK to freeze
impurities
<1.0 EU/μg protein Endotoxin
shipped in
ambient
storage temp.
−20°C
General description
Biochem/physiol Actions
Warning
Physical form
Reconstitution
Other Notes
Ishii, Y., et al. 2008. J. Invest. Dermatol.128, 867.
Kim, K.A., et al. 2008. Mol. Biol. Cell19, 2588.
Binnerts, M.E., et al. 2007. Proc. Natl. Acad. Sci. USA104 14700.
Nam, J.S., et al. 2007. Gene Expr. Patterns7, 306.
Pemberton, T.J., et al. 2007. Dev. Dyn.236, 2245.
Bergmann, C., et al. 2006. Am. J. Hum. Genet.79, 1105.
Blaydon, D.C., et al. 2006. Nat. Genet.38, 1245.
Nam, J.S., et al. 2006. J. Biol. Chem.281, 13247.
Legal Information
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service