Skip to Content
Merck
All Photos(1)

Key Documents

529605-M

Millipore

PP1 Analog

InSolution, ≥97%, ATP-competitive, inhibitor of Src-family tyrosine kinases

Synonym(s):

InSolution PP1 Analog

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H19N5
CAS Number:
Molecular Weight:
317.39
UNSPSC Code:
12352200

Assay

≥97% (HPLC)

form

liquid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles
desiccated (hygroscopic)
protect from light

storage temp.

−70°C

InChI

1S/C19H19N5/c1-19(2,3)24-18-15(17(20)21-11-22-18)16(23-24)14-10-6-8-12-7-4-5-9-13(12)14/h4-11H,1-3H3,(H2,20,21,22)

InChI key

XSHQBIXMLULFEV-UHFFFAOYSA-N

General description

A potent, reversible, ATP-competitive, and cell-permeable inhibitor of Src-family tyrosine kinases that is >800 times more selective for the I338G mutant v-Src (IC50 = 1.5 nM) compared to wild-type v-Src (IC50 = 1.0 µM). Also inhibits wild-type Fyn (IC50 = 600 nM). A 10 mM (1 mg/315 µl) solution of PP1 Analog (Cat. No. 529579) in DMSO.

Warning

Toxicity: Irritant (B)

Physical form

A 10 mM (1 mg/315 µl) solution of PP1 Analog (Cat. No. 529579) in DMSO.

Reconstitution

Following initial thaw, aliquot and freeze (-70°C).

Other Notes

Bishop, A.C., et al. 2000. Nature407, 395.
Bishop, A.C., et al. 1999 J. Am. Chem. Soc.121, 627.

Legal Information

Not available for sale in the United States.
CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

188.6 °F - closed cup - (Dimethylsulfoxide)

Flash Point(C)

87 °C - closed cup - (Dimethylsulfoxide)


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service