Skip to Content
Merck
All Photos(1)

Documents

528178

Sigma-Aldrich

Plasminogen, EACA- and Lysine-Free, Human Plasma

Synonym(s):

Plasminogen, EACA- and Lysine-Free, Human Plasma, Profibrinolysin, Plasma Trypsinogen

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.25

form

lyophilized

Quality Level

specific activity

7 units/mg protein

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

solubility

water: soluble

shipped in

ambient

storage temp.

−20°C

General description

Native plasminogen (EACA- and lysine-free) from human plasma. Plasminogen is a single chain glycoprotein found in normal plasma at ~12 mg per deciliter, and is the inactive precursor of the serine protease plasmin.
Native plasminogen (EACA- and lysine-free) from human plasma. Single chain glycoprotein containing 790 amino acid residues. Found in normal plasma at about 12 mg/100 ml. Inactive precursor of the serine protease plasmin. It is converted to the active protease by cleavage at Arg560.

Warning

Toxicity: Irritant (B)

Unit Definition

One unit is defined as the amount of enzyme that will hydrolyze 1 µmol of N-tosyl-Gly-Pro-Lys-pNA per min at 25°C, pH 7.8.

Physical form

Lyophilized from 20 mM sodium phosphate buffer, 10 mg D-mannitol, 10 mg NaCl.

Preparation Note

Prepared from plasma that has been shown by certified tests to be negative for HBsAg and for antibodies to HIV and HCV.

Reconstitution

Following reconstitution, aliquot and freeze (-20°C) for long-term storage or refrigerate (4°C) for short-term storage. Stock solutions are stable for at least 1 week at 4°C or for 1-2 months at -20°C.

Other Notes

Sun, Z., et al. 2002. Biochim. Biophys. Acta1596, 182.
Robbins, K.C., et al. 1967. J. Biol. Chem.242, 2333.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service