Skip to Content
Merck
All Photos(1)

Key Documents

47589-M

Millipore

Mitomycin C, Streptomyces caespitosus

Synonym(s):

Mitomycin C, Streptomyces caespitosus, Mitocin C, MMC

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H18N4O5
CAS Number:
Molecular Weight:
334.33
MDL number:
UNSPSC Code:
12352200
Pricing and availability is not currently available.

Assay

≥95% (HPLC)

Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

blue-violet

solubility

water: 1 mg/mL

storage temp.

2-8°C

InChI

1S/C15H18N4O5/c1-5-9(16)12(21)8-6(4-24-14(17)22)15(23-2)13-7(18-13)3-19(15)10(8)11(5)20/h6-7,13,18H,3-4,16H2,1-2H3,(H2,17,22)/t6-,7+,13+,15-/m1/s1

InChI key

NWIBSHFKIJFRCO-WUDYKRTCSA-N

General description

Antibiotic and carcinostatic agent. Inhibits DNA synthesis by cross-linking DNA at guanine and adenine residues; disrupts base pairing. Forms covalent adducts into deoxyguanine and guanine under acidic conditions. Induces apoptosis in gastric cancer cells. Blocks the cell cycle at the G2 phase.

Warning

Toxicity: Highly Toxic & Carcinogenic / Teratogenic (I)

Physical form

Provided as 2 mg antibiotic and 48 mg NaCl.

Reconstitution

Following reconstitution, store in the refrigerator (4°C) for short-term storage and in a freezer (-20°C) for longer term. Stock solutions are stable for up to 2 weeks at 4°C and up to 3 months at -20°C.

Other Notes

Kruyt, F.A., et al. 1996. Blood 89, 938.
Schwartz, G.K., et al. 1995. J. Natl. Cancer Inst.87, 1394.
Folnan, R.S. 1993. Oncology 50 (Suppl. 1), 24.
Kawasaki, S., et al. 1992. Jpn. J. Cancer Res.83, 899.
Malkinson, A.M., et al. 1992. Cancer Res.52, 4752.
McEvoy, J.L., et al. 1992. J. Bacteriol.174, 5471.
Tomasz, M., et al. 1987. Science235, 1204.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service