Skip to Content
Merck
All Photos(1)

Documents

420361

Sigma-Aldrich

KMO Inhibitor II, JM6

The KMO Inhibitor II, JM6 controls the biological activity of KMO. This small molecule/inhibitor is primarily used for Neuroscience applications.

Synonym(s):

KMO Inhibitor II, JM6, Kynurenine-3-Monooxygenase Inhibitor II, JM6, Kynurenine-3-Hydroxylase Inhibitor II, JM6, 2-(3,4-Dimethoxybenzenesulfonylamino)-4-(3-nitrophenyl)-5-(piperidin-1-yl)methylthiazole, KMO Inhibitor I, Ro 61-8048 Prodrug

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H26N4O6S2
Molecular Weight:
518.61
UNSPSC Code:
12352200
NACRES:
NA.28

Quality Level

Assay

≥97% (HPLC)

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

yellow

solubility

DMSO: 5 mg/mL

shipped in

wet ice

storage temp.

2-8°C

General description

An orally bioavailable in-vivo prodrug of KMO Inhibitor I, Ro 61-8048 (Cat. No. 420360) that is shown to be metabolically unstable and slowly release Ro 61-8048 in blood and act as a neuroprotectant. Blocks KMO peripherally (IC50 = 4 µM) and sustainingly elevates kynurenic acid (KYNA) brain levels, an antagonist of excitatory amino acid receptors. Shown to prevent synaptic loss and behavioral deficits in AD and HD mouse model and increase the survival.

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Campesan, S., et al. 2011. Chem. Biol.21, 961.
Zwilling, D., et al. 2011. Cell145, 863.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service