Skip to Content
Merck
All Photos(1)

Documents

385898

Sigma-Aldrich

Hsp90, Human

Hsp90, Human, is a ~90 kDa heat shock protein that is constitutively expressed and resides in the cytoplasm. It exists as a dimer and binds to several other cellular proteins.

Synonym(s):

Hsp90 chaperone

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.77

Quality Level

Assay

≥90% (SDS-PAGE)

form

liquid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

shipped in

wet ice

storage temp.

−70°C

General description

Native, human Hsp90. A ~90 kDa heat shock protein that is constitutively expressed and resides in the cytoplasm. It exists as a dimer and binds to other cellular proteins such as p60v-src, steroid receptors, heme-regulated protein kinase, actin, and tubulin. Binds to histones with high affinity and induces chromatin condensation.
Native, human Hsp90. An ~90 kDa heat shock protein constitutively expressed and residues in the cytoplasm. It exists as a dimer and binds to other cellular proteins such as p60v-src, steroid receptors, heme-regulated protein kinase, actin, and tubulin. Binds to histones with high affinity and induces chromatin condensation.

Warning

Toxicity: Standard Handling (A)

Physical form

In 100 mM NaCl, 50 mM Tris-HCl, 1 mM DTT, pH 7.5.

Reconstitution

Following initial thaw, aliquot and freeze (-70°C).

Other Notes

Kim, H.R., et al. 1999. J. Biochem. 126, 1025.Schnaider, T., et al. 1999. Life Sci. 65, 2417.Welch, W.J., et al. 1982. J. Biol. Chem. 257, 14949.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service