Skip to Content
Merck
All Photos(1)

Documents

344216

Sigma-Aldrich

Formyl Peptide Receptor-Like 1 Agonist, WKYMVm

The Formyl Peptide Receptor-Like 1 Agonist, WKYMVm controls the biological activity of Formyl Peptide Receptor-Like 1. This small molecule/inhibitor is primarily used for Activators/Inducers applications.

Synonym(s):

Formyl Peptide Receptor-Like 1 Agonist, WKYMVm, H₂N-WKYMV- D-M-CONH₂, FPRL1 Agonist, WKYMVm, H₂N-WKYMV-D-M-CONH₂, FPRL1 Agonist, WKYMVm

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C41H61N9O7S2
Molecular Weight:
856.11
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥95% (HPLC)

form

lyophilized

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)
protect from light

solubility

DMSO: 10 mg/mL

shipped in

wet ice

storage temp.

−20°C

General description

A potent chemoattractant that enhances leukocyte activity via activation of Formyl Peptide Receptor-Like 1 (FPRL1). More poteut than fmlp at inducing neutrophil chemotaxis, neutrophil complement receptor 3. Shown to activate phospholipase C-mediated inositol phosphate formation in human U266 myeloma cells (EC50 = 510 pM) and in U937 and HL60 cells. Has no effect inositide phosphate synthecs in a variety of other cell lines (e.g. NIH3T3, PC12, Daudi, Sp2, Jurkat, H9, Molt-4, SupT-1, K562, and RBL-2H3). Radioiodinatable at the tyrosine residue.
A potent chemoattractant that enhances leukocyte activity via activation of Formyl Peptide Receptor-Like 1 (FPRL1). Shown to activate phospholipase C-mediated inositol phosphate formation in human U266 myeloma cells (EC50 = 510 pM). Also shown to stimulate inositol phosphate formation in U937 and HL60 cells, but has no effect on a variety of other cell lines (e.g. NIH3T3, PC12, Daudi, Sp2, Jurkat, H9, Molt-4, SupT-1, K562, and RBL-2H3). Stimulates phagocyte chemotaxis and [Ca2+] flux, and is more effective than fMLP (Cat. No. 05-22-2500) in generating superoxide in human neutrophils. Radioiodinatable at the tyrosine residue.

Biochem/physiol Actions

Cell permeable: no
EC50 = 510 pM in activating phospholipase C-mediated inositol phosphate formation in human U266 myeloma cells
Primary Target
Formyl Peptide Receptor-Like 1
Product does not compete with ATP.
Reversible: no

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Sequence

H-Trp-Lys-Tyr-Met-Val-D-Met-CONH₂

Physical form

Supplied as a trifluoroacetate salt.

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Bae, Y.S., et al. 2004. J. Immunol.173, 607.
Bae, Y.S., et al. 2003. Mol. Pharmacol.64, 841.
Le, Y., et al. 1999. J. Immunol.163, 6777.
Seo, J.K., et al. 1997. J. Immunol.158, 1895.
Baek, S.H., et al. 1996. J. Biol. Chem.271, 8170.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service