Skip to Content
Merck
All Photos(1)

Documents

324903

Sigma-Aldrich

Epiregulin, Human, Recombinant

Epiregulin, Human, Recombinant, consists of amino acids 63-108 and expressed in E. coli with an N-terminal methionine.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

Quality Level

Assay

≥97% (SDS-PAGE)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

impurities

<1.0 EU/μg protein Endotoxin

shipped in

ambient

storage temp.

−20°C

General description

Recombinant, human epiregulin consisting of amino acids 63-108 and expressed in E. coli with an N-terminal methionine. Epiregulin, expressed primarily in the placenta and macrophages, stimulates fibroblast, smooth muscle cell, and hepatocyte proliferation.

Biochem/physiol Actions

ED₅₀ = 0.125-0.75 ng/ml as measured in a cell proliferation assay using Balb/3T3 mouse embryonic fibroblasts.

Warning

Toxicity: Standard Handling (A)

Physical form

Lyophilized from 0.2 µm sterile-filtered PBS, BSA.

Reconstitution

Following reconstitution refrigerate (4°C) for short-term storage or aliquot and freeze (-20°C) for long-term storage. Stock solutions are stable for up to 1 month at 4°C or for up to 3 months at -20°C.
Reconstitute to 100 µg/ml in sterile PBS containing at least 0.1% human or bovine serum albumin.

Other Notes

Mishre, R., et al. 2002. Am. J. Physiol. Renal. Physiol.283, F1151.
Shirakata, Y., et al. 2000. J. Biol. Chem.275, 5748.
Shelly, M., et al. 1998. J. Biol. Chem.273, 10496.
Das, S.K., et al. 1997. Dev. Biol.190, 178.
Komurasaki, T., et al. 1997. Oncogene15, 2841.
Toyoda, H., et al. 1997. Biochem. J.326, 69.
Toyoda, H., et al. 1995. J. Biol. Chem.270, 7495.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service