Skip to Content
Merck
All Photos(1)

Documents

324839

Sigma-Aldrich

InhibitorSelect EGFR Signaling Pathway Inhibitor Panel

The InhibitorSelect EGFR Signaling Pathway Inhibitor Panel controls the biological activity of InhibitorSelect.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)
protect from light

solubility

DMSO: soluble (supplied)

storage temp.

−20°C

General description

A panel containing 13 potent and selective inhibitors useful for the study of EGFR signaling pathway. This panel contains the following inhibitors and 15 ml anhydrous DMSO for reconstitution:
A panel containing 13 potent and selective inhibitors useful for the study of EGFR signaling pathway. This panel contains the following inhibitors: 1 mg of Akt Inhibitor VIII, Isozyme-Selective, Akt-1/2 (Cat. No. 124018); 1 mg of PD 153035 (Cat. No. 234490); 1 mg of EGFR Inhibitor (Cat. No. 324674); 5 mg of Et-18-OCH3 (Cat. No. 341207); 5 mg of JNK Inhibitor II (Cat. No. 420119); 5 mg of LY 294002 (Cat. N0. 440202); 5 mg of PD 98059 (Cat. No. 513000); 1 mg of PD 168393 (Cat. No. 513033); 1 mg of PP2 (Cat. No. 529573); 100 µg of Rapamycin (Cat. No. 553210); 1 mg of SB 203580 (Cat. No. 559389); 5 mg of AG 490 (Cat. No. 658401) and 1 mg of ZM 336372 (Cat. No. 692000). Also provided is 15 ml of anhydrous DMSO. Supplied with a data sheet.
The InhibitorSelect EGFR Signaling Pathway Inhibitor Panel controls the biological activity of InhibitorSelect.

Application

InhibitorSelect EGFR Signaling Pathway Inhibitor Panel contains 13 potent and selective inhibitors useful for the study of EGFR signaling pathway.

Packaging

Packaged under inert gas

Warning

Toxicity: Multiple Toxicity Values, refer to MSDS (O)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C) each inhibitor. Each inhibitor solution is stable for up to 3 months at -20°C.
Please refer to information provided on the CD for reconstitution information of individual inhibitors.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 4 - Carc. 2 - Repr. 2 - Skin Sens. 1

Storage Class Code

10 - Combustible liquids

Flash Point(F)

188.6 °F

Flash Point(C)

87 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service