Skip to Content
Merck
All Photos(1)

Documents

1.94609

Supelco

Ethylene glycol monomethyl ether

for analysis EMPARTA® ACS

Synonym(s):

2-Methoxyethanol, Ethylene glycol monomethyl ether, Methyl glycol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OCH2CH2OH
CAS Number:
Molecular Weight:
76.09
Beilstein:
1731074
MDL number:
UNSPSC Code:
12352112

grade

ACS reagent

vapor density

2.62 (vs air)

vapor pressure

6.17 mmHg ( 20 °C)

autoignition temp.

548 °F

expl. lim.

24.5 %

refractive index

n20/D 1.402 (lit.)

bp

124-125 °C (lit.)

mp

−85 °C (lit.)

density

0.965 g/mL at 25 °C (lit.)

storage temp.

room temp

SMILES string

COCCO

InChI

1S/C3H8O2/c1-5-3-2-4/h4H,2-3H2,1H3

InChI key

XNWFRZJHXBZDAG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

EMPARTA is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Flam. Liq. 3 - Repr. 1B - STOT RE 2 - STOT SE 1

Target Organs

Immune system, thymus

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

104.0 °F - closed cup

Flash Point(C)

40 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service