Skip to Content
Merck
All Photos(1)

Key Documents

189405-M

Millipore

Aurora Kinase Inhibitor III

The Aurora Kinase Inhibitor III, also referenced under CAS 879127-16-9, controls the biological activity of Aurora Kinase. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

Synonym(s):

Aurora Kinase Inhibitor III, Cyclopropanecarboxylic acid-(3-(4-(3-trifluoromethyl-phenylamino)-pyrimidin-2-ylamino)-phenyl)-amide, Aurora Kinase Inhibitor III

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H18F3N5O
CAS Number:
Molecular Weight:
413.40
MDL number:
UNSPSC Code:
12352200
Pricing and availability is not currently available.

Assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

solubility

ethanol: 20 mg/mL
DMSO: 50 mg/mL

storage temp.

2-8°C

InChI

1S/C21H18F3N5O/c22-21(23,24)14-3-1-4-15(11-14)26-18-9-10-25-20(29-18)28-17-6-2-5-16(12-17)27-19(30)13-7-8-13/h1-6,9-13H,7-8H2,(H,27,30)(H2,25,26,28,29)

InChI key

RDTDWGQDFJPTPD-UHFFFAOYSA-N

General description

A cell-permeable 2,4-dianilinopyrimidine compound that acts as an ATP-competitive, potent, but non-selective inhibitor of Aurora A (IC50 = 42 nM). At higher concentrations, also inhibits the activities of other kinases, such as Lck, Bmx, IGF-1R, and Syk (IC50 = 131, 386, 591, and 887 nM, respectively).

Biochem/physiol Actions

Primary Target
Aurora Kinase A
Target IC50: 42 nM against Aurora A; 131, 386, 591, and 887 nM, against Lck, Bmx, IGF-1R, and Syk , respectively

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Zhang, Q., et al. 2006. J. Am. Chem. Soc.128, 2182.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service