1.66783
Tryptic Soy Agar
with Lecithin, Tween®, Histidine and Sodium thiosulphate, ICR+ Contact plate, irradiated, triple packed, for environmental monitoring (Isolator and Clean room)
Synonym(s):
TSA, Soybean Casein Digest Agar, ICR plus Contact plate
About This Item
Recommended Products
material
polystyrene plate
Quality Level
Agency
EP
USP
sterility
sterile; γ-irradiated
form
plate
feature
2D bar code
ready-to-use
packaging
pkg of (20/200 Plates per box)
D
55 mm
pH
(7.3 (in H2O))
application(s)
microbiology
pharmaceutical
surface monitoring
storage temp.
15-25°C
suitability
bacteria
fungi
General description
Application
Packaging
1667830200-CN: 200 Plates per box
Legal Information
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service