Skip to Content
Merck
All Photos(1)

Documents

1.46770

Millipore

ReadyPlate 55 KIT m Green Agar

suitable for detection, filtration, box of 150 sets 55 mm plates and membrane filters

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106227
NACRES:
NA.74

material

gridded white mixed cellulose esters (MCE) membrane

Quality Level

Agency

ISO

feature

ready-to-use

packaging

box of 150 sets 55 mm plates and membrane filters

manufacturer/tradename

ReadyPlate

pore size

0.45 μm pore size

application(s)

detection
filtration

food and beverages
industrial qc
microbiology
water monitoring

storage temp.

15-25°C

General description

Application: Detection of yeast, mould and aciduric bacteria, Membrane filtration method

Packaging: 55 mm plate and EZ PAK membrane filter (150 sets per box)

Synonym: Schaufuss Pottinger Medium

Legal Information

READYPLATE is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Irrit. 2 - Flam. Sol. 1

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service