Skip to Content
Merck
All Photos(1)

Key Documents

1.46256

Millipore

Standard I Nutrient Agar

Application: Standard media
Packaging: 90 mm plate (20 plates per box)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106217
NACRES:
NA.74
Pricing and availability is not currently available.

Quality Level

form

solid

feature

ready-to-use

pH

7.4 ( in H2O)

application(s)

industrial qc
microbiology
pharmaceutical

storage temp.

2-8°C

suitability

nonselective for

General description

Application: Standard media

Packaging: 90 mm plate (20 plates per box)

Storage Class Code

11 - Combustible Solids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service