Skip to Content
Merck
All Photos(1)

Documents

1.24513

Supelco

Bathophenanthroline

GR for analysis

Synonym(s):

Bathophenanthroline, 4,7-Diphenyl-1,10-phenanthroline

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H16N2
CAS Number:
Molecular Weight:
332.40
MDL number:
UNSPSC Code:
12352005
EC Index Number:
216-767-1

Quality Level

form

solid

mp

215-220 °C

bulk density

400 kg/m3

storage temp.

15-25°C

InChI

1S/C24H16N2/c1-3-7-17(8-4-1)19-13-15-25-23-21(19)11-12-22-20(14-16-26-24(22)23)18-9-5-2-6-10-18/h1-16H

InChI key

DHDHJYNTEFLIHY-UHFFFAOYSA-N

Analysis Note

Assay (perchloric acid titration): ≥ 99 %
Identity (IR-spectrum): passes test
Melting range: 217 - 222 °C

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service