Skip to Content
Merck

1.09828

SAFC

di-Potassium hydrogen phosphate anhydrous

low in sodium, EMPROVE® API, Ph. Eur., BP, USP

Pharma Manufacturing

Synonym(s):

Potassium phosphate dibasic, Dipotassium Phosphate, Dipotassium hydrogen phosphate, Dipotassium hydrogenphosphate, Dipotassium phosphate, sec.-Potassium phosphate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
K2HPO4
CAS Number:
Molecular Weight:
174.18
MDL number:
UNSPSC Code:
12352302
EC Index Number:
 231-834-5

biological source

synthetic

Quality Level

grade

GMP
Halal
Kosher

Agency

BP
Ph. Eur.
USP

product line

EMPROVE® API

form

crystalline

impurities

low in endotoxins

application(s)

clinical nutrition
enterals
parenterals
pharmaceutical

storage temp.

2-25°C

SMILES string

[K+].[K+].OP([O-])([O-])=O

InChI

1S/2K.H3O4P/c;;1-5(2,3)4/h;;(H3,1,2,3,4)/q2*+1;/p-2

InChI key

ZPWVASYFFYYZEW-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

We provide industry-leading production know-how and comprehensive regulatory support for high-quality APIs, backed by nearly 200 years of experience. From a complete portfolio of innovative folates to custom synthesis, our customized and scalable solutions meet your specific production needs.
As part of our EMPROVE® Program, our raw materials are offered with extensive documentation facilitating compliance of your pharma and biopharma product, full supply chain transparency and risk mitigation. Our SAFC® portfolio of high-quality products for biopharmaceutical and pharmaceutical formulation and production withstands strict quality control procedures and is produced according to applicable cGMP guidelines.

Application

di-Potassium hydrogen phosphate as active pharmaceutical ingredients is used in clinical nutrition (enteral and parenteral) and for electrolyte replenishment.

Packaging

1.09828.0001 // 3x 1kg // PE bottles (in carton)
1.09828.1000 // 1kg // PE bottle
1.09828.9013 // 12kg // PE bag (in PE pail)
1.09828.9027 // 25kg // PE bag (in PE drum)

Legal Information

Emprove is a registered trademark of Merck KGaA, Darmstadt, Germany
SAFC is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service