Skip to Content
Merck
All Photos(1)

Documents

878101P

Avanti

C17 Lyso PAF

Avanti Research - A Croda Brand 878101P, powder

Synonym(s):

1-O-heptadecyl-2-hydroxy-sn-glycero-3-phosphocholine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H54NO6P
CAS Number:
Molecular Weight:
495.67
UNSPSC Code:
12352211
NACRES:
NA.25

form

powder

packaging

pkg of 1 × 10 mg (878101P-10mg)

manufacturer/tradename

Avanti Research - A Croda Brand 878101P

lipid type

bioactive lipids
phosphoglycerides

shipped in

dry ice

storage temp.

−20°C

SMILES string

O[C@](COP([O-])(OCC[N+](C)(C)C)=O)([H])COCCCCCCCCCCCCCCCCC

General description

C17 Lyso PAF, also known as 1-O-heptadecyl-2-hydroxy-sn-glycero-3-phosphocholine, is a precursor and metabolite of 1-O-heptadecyl-2-acetyl-sn-glycero-3-phosphocholine (C17:0 PAF). Lyso-platelet activating factor (PAF) is synthesized from PAF acetylhydrolases by the degradation of PAF. It is also synthesized from 1-O-alkyl-2-arachidonoyl-sn-glycero-3-phosphocholine (1-alkyl-phosphatidylcholine; PC) by the enzyme phospholipase A2 via the remodeling pathway.

Packaging

5 mL Amber Glass Screw Cap Vial (878101P-10mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hideo Shindou et al.
The Journal of biological chemistry, 282(9), 6532-6539 (2006-12-22)
Platelet-activating factor (PAF) is a potent proinflammatory lipid mediator eliciting a variety of cellular functions. Lipid mediators, including PAF are produced from membrane phospholipids by enzymatic cascades. Although a G protein-coupled PAF receptor and degradation enzymes have been cloned and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service