Z743886
ART® SoftFit L Reload System
HLT 200L, size 200 μL, reload insert, non-sterile
Synonym(s):
aerosol barrier tips, barrier filter pipette tip, barrier filter tips, barrier tip, filter pipette tip, filter tip, pipet tip
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
sterility
non-sterile
feature
HLT 200L
reload insert
packaging
pkg of 5X960 ea
manufacturer/tradename
Molecular BioProducts 3752-RI
technique(s)
PCR: suitable
size
200 μL
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Attain the accuracy and precision you anticipate from your pipette when pipetting samples, each time utilizing “SoftFit-L”pipette tips. This distinctive system offers preloaded pipette tips encased within volume-sized color-coded inserts, complemented by the choice of a user-friendly "push down" reload pack.
Features and Benefits
- Reload towers are an eco-friendly packaging choice, effectively reducing plastic waste and essential storage area.
- Designed to match with empty SoftFit-L racks.
- Color-coded inserts enable quick recognition of tip volume.
- Free of DNase, RNase, DNA, ATP, and endotoxin.
Legal Information
ART is a registered trademark of Molecular BioProducts, Inc.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service