Skip to Content
Merck
All Photos(1)

Documents

W434900

Sigma-Aldrich

cis-4-Decen-1-ol

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)4CH=CH(CH2)3OH
CAS Number:
Molecular Weight:
156.27
FEMA Number:
4349
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:

biological source

synthetic

Assay

97%

refractive index

n20/D 1.452 (lit.)

bp

231-232 °C (lit.)

density

0.853 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Organoleptic

fruity

SMILES string

[H]\C(CCCO)=C(/[H])CCCCC

InChI

1S/C10H20O/c1-2-3-4-5-6-7-8-9-10-11/h6-7,11H,2-5,8-10H2,1H3/b7-6-

InChI key

VUNFOJWKJSYIDH-SREVYHEPSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

>230.0 °F

Flash Point(C)

> 110 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service