Skip to Content
Merck
All Photos(1)

Documents

W354600

Sigma-Aldrich

5-Ethyl-2-methylpyridine

≥96%

Synonym(s):

5-Ethyl-2-picoline, NSC 1984

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H11N
CAS Number:
Molecular Weight:
121.18
FEMA Number:
3546
Beilstein:
109269
EC Number:
Council of Europe no.:
11385
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
14.066
NACRES:
NA.21

biological source

synthetic

vapor density

4.2 (vs air)

Assay

≥96%

refractive index

n20/D 1.497 (lit.)

bp

178 °C (lit.)

density

0.919 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

nutty; earthy; roasted

SMILES string

CCc1ccc(C)nc1

InChI

1S/C8H11N/c1-3-8-5-4-7(2)9-6-8/h4-6H,3H2,1-2H3

InChI key

NTSLROIKFLNUIJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Masaaki Fujikawa et al.
Bioorganic & medicinal chemistry, 15(11), 3756-3767 (2007-04-10)
We previously reported a classical quantitative structure-activity relationship (QSAR) equation for permeability coefficients (P(app-pampa)) by parallel artificial membrane permeation assay (PAMPA) of structurally diverse compounds with simple physicochemical parameters, hydrophobicity at a particular pH (logP(oct) and |pK(a)-pH|), hydrogen-accepting ability (SA(HA))

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service