Skip to Content
Merck
All Photos(1)

Documents

SIK7719-10

Sigma-Aldrich

1-Methyl-1,2,4-(dodecylphosphonic acid)triazolium bromide

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H31BrN3O3P
CAS Number:
Molecular Weight:
412.30
UNSPSC Code:
12352106
NACRES:
NA.23

Assay

97%

form

powder

shelf life

3 years under the recommended conditions

color

white

Application

Quaternary ammonium compounds (QACs) are low toxicity compounds that form effective protective layers against biodeterioration. They are widely known to be bioactive and have a high antimicrobial activity. They can be used to create positively charged surfaces with antimicrobial properties.

Legal Information

Product of SiKEMIA

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service