All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C34H46N4O7S
CAS Number:
Molecular Weight:
654.82
UNSPSC Code:
12352200
PubChem Substance ID:
Recommended Products
Assay
>90%
form
solid or viscous liquid
reaction suitability
reagent type: cross-linking reagent
shipped in
ambient
storage temp.
−20°C
SMILES string
O=C(NCCCOCCOCCOCCCNC(C1=CC=C(C=C1)C(C2=CC=CC=C2)=O)=O)CCCCC3C4NC(NC4CS3)=O
Features and Benefits
Biotin-dPEG®3-benzophenone combines biotin with a 16.9 Å hydrophilic spacer and a highly photoreactive group to permit biotin labeling of molecules that do not have an easily useable functionality (for example, polymers). In one of the most efficient photoreactive conjugation mechanisms available today, benzophenone is activated by 365 nm UV light to a highly reactive triplet state ketone that inserts covalently into CH and NH structures. This is a unique product that is highly useful for certain types of molecules that customers may want to biotinylate.
Legal Information
Products Protected under U.S. Patents # 7,888,536 B2
dPEG is a registered trademark of Quanta BioDesign
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service