Skip to Content
Merck
All Photos(1)

Documents

OCNQCK620

Sigma-Aldrich

EDC Conjugation Kit for Carboxylic Cd-based Core/Shell Quantum Dots

fluorescence λem 620 nm, for 4 conjugation

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
43211915
NACRES:
NA.51

fluorescence

λem 620 nm

Looking for similar products? Visit Product Comparison Guide

General description

Components:
  • QSH
  • EDC
  • Coupling Buffer
  • Quenching Buffer
  • Storage Buffer
  • Tubes 4 x 1.7 mL

Legal Information

Product of Ocean Nanotech, LLC.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT RE 2 Oral

Target Organs

Stomach,large intestine,lymph node

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Cadmium-free quantum dots for bioimaging offer design and synthesis strategies, enhancing bioimaging and sensing applications.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service