Skip to Content
Merck
All Photos(1)

Documents

MT30046249

Mettler Toledo SevenExcellence pH/Ion meter S500

S500 standard kit, with InLab® Expert Pro-ISM, AC/DC input 230 V AC, universal plug set

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115603
NACRES:
NB.61

description

Concentration range 0.001 … 999 999 mg/L, ppm ; 0.0001 … 100 mol/L, % ; 0.001 … 100 000 mmol/L (+/- 0.5%)

feature

with InLab® Expert Pro-ISM

manufacturer/tradename

Mettler-Toledo 30046249

mV measuring range

-2000-2000, accuracy: ±1

temperature measuring range

-30-130 °C, accuracy: ±0.1

pH range

-2-20

Looking for similar products? Visit Product Comparison Guide

General description

Includes meter with a pH/Ion expansion unit, 2 blank expansion units, InLab® Expert Pro-ISM, guide to pH-measurement, guide to Ion measurement and calibration sachets
Mettler Toledo SevenExcellence pH/Ion meter – intuitive touchscreen operation and high measurement performance combined with a complete security package for safe results and solid compliance. This single channel instrument is modularly expandable with additional measurement parameters at any time!

  • Methods for incremental ion measurements, guiding the user through the individual steps
  • Large 7 inch color display and intuitive menu guidance in 10 languages
  • Flexible method concept for high reproducibility and security
  • Extra security thanks to sophisticated user management and Intelligent Sensor Management (ISM®)
  • Integrated interfaces (USB, RS232 and Ethernet) for data exchange
  • Comprehensive service package including IQ/OQ/PQ

Included in every SevenExcellence:
uPlace electrode holder with a perfect vertical motion. It can be operated with one hand and used either freestanding or attached to the left or right side of the instrument.
LabX direct pH PC software for transferring real-time measurement data and archiving your results

Legal Information

ISM is a registered trademark of Mettler-Toledo, Inc.
InLab is a registered trademark of Mettler-Toledo, Inc.
SevenExcellence is a trademark of Mettler-Toledo, Inc.
uPlace is a trademark of Mettler-Toledo, Inc.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service