Skip to Content
Merck
All Photos(1)

Key Documents

H26502

Sigma-Aldrich

N-(2-Hydroxyethyl)ethylenediamine-N,N′,N′-triacetic acid

99%

Synonym(s):

N-Carboxymethyl-N′-(2-hydroxyethyl)-N,N′-ethylenediglycine, HEDTA, HEEDTA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOCH2CH2N(CH2CO2H)CH2CH2N(CH2CO2H)2
CAS Number:
Molecular Weight:
278.26
Beilstein:
1804795
EC Number:
MDL number:
UNSPSC Code:
12352100

Assay

99%

mp

212 °C (dec.) (lit.)

SMILES string

OCCN(CCN(CC(O)=O)CC(O)=O)CC(O)=O

InChI

1S/C10H18N2O7/c13-4-3-11(5-8(14)15)1-2-12(6-9(16)17)7-10(18)19/h13H,1-7H2,(H,14,15)(H,16,17)(H,18,19)

InChI key

URDCARMUOSMFFI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service