Skip to Content
Merck
All Photos(2)

Key Documents

D0261

Sigma-Aldrich

N-(2,4-Dinitrophenyl) taurine sodium salt

Synonym(s):

DNP-Taurine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C8H8N3O7SNa
CAS Number:
Molecular Weight:
313.22
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

form

solid

storage temp.

−20°C

SMILES string

[Na].OS(=O)(=O)CCNc1ccc(cc1N(=O)=O)N(=O)=O

InChI

1S/C8H9N3O7S.Na/c12-10(13)6-1-2-7(8(5-6)11(14)15)9-3-4-19(16,17)18;/h1-2,5,9H,3-4H2,(H,16,17,18);/q;+1/p-1

InChI key

AXJWYGAWAZMPCM-UHFFFAOYSA-M

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service