Skip to Content
Merck
All Photos(1)

Key Documents

CVT00101

Sigma-Aldrich

Ethyl 5-(2-methoxyphenyl)-1-(2,2,2-trifluoroethyl)-1H-pyrazole-4-carboxylate

Synonym(s):

5-(2-Methoxyphenyl)-1-(2,2,2-trifluoroethyl)-1H-pyrazole-4-carboxylic acid ethyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H15F3N2O3
Molecular Weight:
328.29
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

form

solid

SMILES string

O=C(OCC)C1=C(N(CC(F)(F)F)N=C1)C2=CC=CC=C2OC

InChI

1S/C15H15F3N2O3/c1-3-23-14(21)11-8-19-20(9-15(16,17)18)13(11)10-6-4-5-7-12(10)22-2/h4-8H,3,9H2,1-2H3

InChI key

LQBPSWFLPWRLLI-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service