Skip to Content
Merck
All Photos(1)

Documents

CLS7095B5X

Corning® Pasteur pipettes, non-sterile

L 5 3/4 in. (146 mm), standard tip, soda lime

Synonym(s):

pipettes, serological pipets, serological pipettes

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121500
eCl@ss:
32079210
NACRES:
NB.04

material

borosilicate glass

sterility

non-sterile

feature

graduations: no
subdivision: no

packaging

case of 1000 ea
pack of 200 ea

manufacturer/tradename

Corning 7095B-5X

L

5 3/4 in. (146 mm)

Looking for similar products? Visit Product Comparison Guide

General description

  • Corning® disposable Pasteur pipettes are made of sturdy soda-lime glass.
  • Single-use pipettes prevent the risk of cross-contamination of valuable specimens or laboratory reagents.
  • Feature a constriction one inch below the top to facilitate plugging.
  • Ideal for use in non-volumetric transfer work in bacteriology, immunology, hematology, and serology studies, as well as blood bank and clinical chemistry procedures.

Legal Information

Corning is a registered trademark of Corning, Inc.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

  1. Which document(s) contains shelf-life or expiration date information for a given product?

    If available for a given product, the recommended re-test date or the expiration date can be found on the Certificate of Analysis.

  2. How do I get lot-specific information or a Certificate of Analysis?

    The lot specific COA document can be found by entering the lot number above under the "Documents" section.

  3. How do I find price and availability?

    There are several ways to find pricing and availability for our products. Once you log onto our website, you will find the price and availability displayed on the product detail page. You can contact any of our Customer Sales and Service offices to receive a quote.  USA customers:  1-800-325-3010 or view local office numbers.

  4. What is the Department of Transportation shipping information for this product?

    Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product. 

  5. What is the outer diameter of product CLS7095B5X (Corning® Pasteur pipettes, non-sterile)?

    Product CLS7095B5X (Corning® Pasteur pipettes, non-sterile) has an outer diameter of 6.5 mm.

  6. My question is not addressed here, how can I contact Technical Service for assistance?

    Ask a Scientist here.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service