Skip to Content
Merck
All Photos(1)

Key Documents

C25805

Sigma-Aldrich

4-Chlorobenzotrichloride

98%

Synonym(s):

α,α,α,4-Tetrachlorotoluene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClC6H4CCl3
CAS Number:
Molecular Weight:
229.92
Beilstein:
1866549
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

98%

refractive index

n20/D 1.572 (lit.)

bp

245 °C (lit.)

density

1.495 g/mL at 25 °C (lit.)

SMILES string

Clc1ccc(cc1)C(Cl)(Cl)Cl

Looking for similar products? Visit Product Comparison Guide

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Carc. 1B - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

>235.4 °F - closed cup

Flash Point(C)

> 113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

[Determination of the MPEL for alpha, alpha, alpha-trichloro-4-chlorotoluene in the air of work areas].
A I Khalepo et al.
Gigiena truda i professional'nye zabolevaniia, (6)(6), 41-43 (1984-06-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service