BAF196140000
SP Bel-Art® Wye (Y) Tubing Connectors
tubing I.D. 0.25 in. (6 mm)
Synonym(s):
connectors, tubing, tubing coupler
All Photos(2)
About This Item
Recommended Products
packaging
pack of 12 ea
manufacturer/tradename
Bel-Art F19614-0000
L × W
1.3125 in. (33.3 mm) × 2.0625 in. (52.8 mm)
tubing I.D.
0.25 in. (6 mm)
General description
Use these fittings to ensure a tight, secure connection between lengths of tubing having the same inner diameter (I.D.).
Features and Benefits
- Multiple barbs per arm provide additional contact between the fitting and the tubing for extra gripping power
- Chemical-resistant polypropylene plastic for use with a wide range of liquids or gases
- FDA-grade for direct contact with food, drugs, and cosmetics
- Autoclavable
Legal Information
Bel-Art is a registered trademark of SP Scienceware, a division of SP