B60002
[(1R)-(endo,anti)]-(+)-3-Bromocamphor-8-sulfonic acid ammonium salt
99%
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C10H15BrO4S · NH3
CAS Number:
Molecular Weight:
328.22
EC Number:
MDL number:
UNSPSC Code:
12352115
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
99%
form
powder
optical activity
[α]25/D +84.5°, c = 4 in H2O
mp
284 °C (dec.) (lit.)
SMILES string
N.C[C@@]1(CS(O)(=O)=O)[C@@H]2CC[C@@]1(C)C(=O)[C@H]2Br
InChI
1S/C10H15BrO4S.H3N/c1-9-4-3-6(7(11)8(9)12)10(9,2)5-16(13,14)15;/h6-7H,3-5H2,1-2H3,(H,13,14,15);1H3/t6-,7+,9+,10-;/m1./s1
InChI key
GFBVBBRNPGPROZ-ZEMXJPTRSA-N
Application
[(1R)-(endo,anti)]-(+)-3-Bromocamphor-8-sulfonic acid ammonium salt can be used as a chiral resolving reagent in the resolution of piperidine intermediate, which is used in the synthesis of ORL-1 antagonist named J-113397.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
A new synthesis of the ORL-1 antagonist 1-[(3R, 4R)-1-cyclooctylmethyl-3-hydroxymethyl-4-piperidinyl]-3-ethyl-1, 3-dihydro-2H-benzimidazol-2-one (J-113397) and activity in a calcium mobilization assay
Smith ED, et al.
Bioorganic & Medicinal Chemistry, 16(2), 822-829 (2008)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service