All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C7H6FNO4S
CAS Number:
Molecular Weight:
219.19
UNSPSC Code:
12352100
Assay
95%
form
liquid
reaction suitability
reaction type: click chemistry
refractive index
n/D 1.538
density
1.467 g/L at 25 °C
InChI
1S/C7H6FNO4S/c1-5-2-3-6(9(10)11)4-7(5)14(8,12)13/h2-4H,1H3
InChI key
DVIHYWIICFLCGC-UHFFFAOYSA-N
Application
Sulfonyl fluoride motif can be used as a connector for the assembly of -SO2- linked small molecules with proteins or nucleic acids. This new click chemistry approach through sulfates is a complimentary approach to using amides and phosphate groups as linkers.
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Skin Corr. 1B
Storage Class Code
8A - Combustible corrosive hazardous materials
WGK
WGK 3
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service