Skip to Content
Merck
All Photos(1)

Key Documents

96742

Sigma-Aldrich

Z-Leu-ONp

≥98.0% (TLC)

Synonym(s):

Z-L-leucine 4-nitrophenyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H22N2O6
CAS Number:
Molecular Weight:
386.40
Beilstein:
1896374
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

≥98.0% (TLC)

storage temp.

2-8°C

SMILES string

CC(C)C[C@H](NC(=O)OCc1ccccc1)C(=O)Oc2ccc(cc2)[N+]([O-])=O


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service