Skip to Content
Merck
All Photos(1)

Key Documents

96087

Sigma-Aldrich

N-Z-1,6-hexanediamine hydrochloride

≥97% (AT)

Synonym(s):

Benzyl N-(6-aminohexyl)carbamate hydrochloride, N-Z-1,6-diaminohexane hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH2OCONH(CH2)6NH2 · HCl
CAS Number:
Molecular Weight:
286.80
Beilstein:
4727129
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22

Assay

≥97% (AT)

form

crystals

reaction suitability

reagent type: cross-linking reagent

mp

175-180 °C

SMILES string

Cl[H].NCCCCCCNC(=O)OCc1ccccc1

InChI

1S/C14H22N2O2.ClH/c15-10-6-1-2-7-11-16-14(17)18-12-13-8-4-3-5-9-13;/h3-5,8-9H,1-2,6-7,10-12,15H2,(H,16,17);1H

InChI key

GMSZGOGHNFZOMF-UHFFFAOYSA-N

Other Notes

Introduction of a hexamethylenediamine linker

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M M Ponpipom et al.
Carbohydrate research, 113(1), 45-56 (1983-02-16)
Spacer arms 2.1-3.7 nm (21-37 A) long were prepared, and coupled with the methyl beta-glycoside of N-acetylmuramyl-L-alanyl-D-isoglutamine benzyl ester, to give blocked 6-acylates. Deprotection was effected with palladium chloride and triethyl-silane. Chemical conjugates of MDP-meningococcal group C polysaccharide were then

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service