Skip to Content
Merck
All Photos(1)

Key Documents

95421

Sigma-Aldrich

Bis[tris(4-(heptadecafluorooctyl)phenyl)phosphine]palladium(II) dichloride

≥95.0% (31P-NMR)

Synonym(s):

Bis[tris(4-(perfluorooctyl)phenyl)phosphine]palladium(II) dichloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C84H24Cl2F102P2Pd
CAS Number:
Molecular Weight:
3210.20
MDL number:
UNSPSC Code:
12352200

Assay

≥95.0% (31P-NMR)

Application

Catalyst for Stille and Suzuki couplings in "Fluorous Biphase Systems" (FBS)

Other Notes

Perfluoro-tagged Pd complexes as catalysts for Stille couplings in a fluorous biphase system, palladium-catalyzed Suzuki couplings.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Dam. 1 - Lact. - Repr. 1B - STOT RE 1

Target Organs

Liver

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T. Osswald et al.
Tetrahedron Letters, 42, 2965-2965 (2001)
S. Schneider et al.
Helvetica Chimica Acta, 84, 735-735 (2001)
S. Schneider et al.
Angewandte Chemie (International Edition in English), 112, 4293-4293 (2000)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service