Skip to Content
Merck
All Photos(1)

Key Documents

924350

Tris(1-adamantyl)phosphine

Umicore

Synonym(s):

P(Ad)3

Sign Into View Organizational & Contract Pricing


About This Item

Quality Level

form

powder

reaction suitability

reagent type: ligand

functional group

phosphine

Application

Tris(1-adamantyl)phosphine, P(Ad)3, is a crystalline solid phosphine ligand that exhibits electron releasing character greater than most alkylphosphines. This ligand had demonstrated uses in many cross-coupling reactions, including the Suzuki-Miyaura coupling.

Storage and Stability

Do not store above 25°C (77°F). Store in a tightly closed original container under dry inert gas, protected from direct sunlight in a dry, cool and well-ventilated area.

Legal Information

Product of Umicore

This product, its manufacturing or use, is the subject of one or more issued or pending U.S. Patents (and foreign equivalents) owned or controlled by Umicore PMC. The purchase of this product from Umicore PMC through Sigma-Aldrich, its affiliates or their authorized distributors conveys to the buyer a limited, one-time, non-exclusive, non-transferable, non-assignable license. Buyer′s use of this product may infringe patents owned or controlled by third parties. It is the sole responsibility of buyer to ensure that its use of the product does not infringe the patent rights of third parties or exceed the scope of the license granted herein.

Additional information available at www.pmc.umicore.com

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tri(1-adamantyl)phosphine: Expanding the Boundary of Electron-Releasing Character Available to Organophosphorus Compounds
Liye Chen, Peng Ren, and Brad P. Carrow
Journal of the American Chemical Society, Volume 138, Issue 20, 6392-6395 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service