Skip to Content
Merck
All Photos(1)

Key Documents

900588

Sigma-Aldrich

(S)-2-(Piperidin-2-yl)pyridine

95%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H14N2
CAS Number:
Molecular Weight:
162.23
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.22
form:
liquid
Assay:
95%
Pricing and availability is not currently available.

Assay

95%

form

liquid

storage temp.

2-8°C

InChI

1S/C10H14N2/c1-3-7-11-9(5-1)10-6-2-4-8-12-10/h1,3,5,7,10,12H,2,4,6,8H2/t10-/m0/s1

InChI key

KXRQQPIHUMSJSS-JTQLQIEISA-N

Application

Chiral building block developed using Liverpool ChiroChem-patented technology.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service