Skip to Content
Merck
All Photos(1)

Key Documents

858587

Sigma-Aldrich

Phosphoenolpyruvic acid tri(cyclohexylamine) salt monohydrate

98%

Synonym(s):

PEP, Tri(cyclohexylamine) salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H44N3O6P · H2O
CAS Number:
Molecular Weight:
483.58
MDL number:
UNSPSC Code:
12352106

Assay

98%

SMILES string

O.NC1CCCCC1.NC2CCCCC2.NC3CCCCC3.OC(=O)C(=C)OP(O)(O)=O

Application

Substrate for many kinase reactions.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anneli Borg et al.
The Journal of biological chemistry, 290(6), 3440-3454 (2014-12-03)
The antibiotic fusidic acid (FA) targets elongation factor G (EF-G) and inhibits ribosomal peptide elongation and ribosome recycling, but deeper mechanistic aspects of FA action have remained unknown. Using quench flow and stopped flow experiments in a biochemical system for
Yousuke Nakai et al.
Gastrointestinal endoscopy, 81(1), 119-126 (2014-12-03)
Wire-guided cannulation (WGC) was reported to decrease post-ERCP pancreatitis (PEP), but risk factors for PEP in WGC are not fully elucidated. To evaluate the incidence and risk factors of PEP in WGC. Single-center retrospective study. Academic center. A total of
Neel S Choksi et al.
Gastrointestinal endoscopy, 81(1), 150-155 (2014-12-21)
It is believed, based on limited observational data, that an unsuccessful attempt to place a prophylactic pancreatic stent substantially increases the risk of post-ERCP pancreatitis (PEP). To better understand the risk of PEP in patients with failed pancreatic stent placement
Peter Schwandt et al.
American journal of hypertension, 28(5), 672-679 (2014-11-22)
Strong associations between blood pressure (BP) and overweight raise the question whether overweight children (body mass index (BMI) ≥85th percentile) should be included in the normative database. Using the LMS (Lamda-Mu-Sigma) method, we developed age-, gender-, and height-adjusted percentile curves
David Nicolás-Pérez et al.
Pancreas, 44(2), 204-210 (2014-11-20)
The aim of the present study was to perform a comparative cost-effectiveness analysis of the different strategies used to prevent post-endoscopic retrograde cholangiopancreatography (ERCP) acute pancreatitis. We performed a cost-effectiveness decision analysis of 4 prophylactic strategies (nonsteroidal anti-inflammatory drugs or

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service